Five-Channel Timer, Sper Scientific
Supplier:   Sper Scientific
Five-Channel Timer, Sper Scientific
Supplier:  Sper Scientific
Certificates
Five-Channel Timer, Sper Scientific
Five-Channel Timer, Sper Scientific
2 Options Available Below
Product Details & Documents
This digital count down/count up timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours, plus clock button.
  • Extra large display indicates channels in use
  • Each one has a distinctive 1 minute alarm
Automatically begins to count up to indicate time elapsed after count down reaches zero. Memory recalls last timer setting or reuse. Also has a clock mode. Has a stand, pocket clip, and magnet.
 : N.I.S.T. traceable certificate of compliance for the certified option.
Product Family Options
Product Information
  • Désignation
    W×H×D
    Weight
    Availability
    Price
  • 5 channel timer
    2⅞×2½×½"
    2 oz.
    Log in for availability
    Log in for availability
    Each
    Each - $144.92
    $144.92
    New Customer Promotional Price
    Each
       $144.92
    Supplier Number: 810015
  • 5 channel timer, certified
    2⅞×2½×½"
    2 oz.
    Log in for availability
    Log in for availability
    Each
    Each - $283.36
    $283.36
    Each
       $283.36
    Supplier Number: 810015C