WorkWipes® Reclaimed Colored Healthcare Linen in Bag, New Pig
Supplier: New Pig
Certificates
About this item
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Low-lint, laundered material is ideal for staining, wiping, polishing and other general tasks
Eco- and cost-friendly rags are made from post-consumer recycled cotton and cotton blend healthcare gowns, scrubs and sheets.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Scanned for metal. Not sorted for printing. Washed.
Frequently Bought Together