WorkWipes® Reclaimed Colored T-Shirt in Bag, New Pig
Fournisseur: New Pig
Certificats
À propos de cet article
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Durable, laundered, low-lint material is ideal for staining, wiping, polishing and other general-use tasks
Eco- and cost-friendly rags are made from post-consumer recycled cotton and cotton blend T-shirts, so they're strong and absorbent.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Ordering information: Scanned for metal. Sorted for printing. Not washed.
Frequently Bought Together