Project Timer, Sper Scientific
Project Timer, Sper Scientific
Supplier:   Sper Scientific
Project Timer, Sper Scientific
Supplier:  Sper Scientific
Certificates
Project Timer, Sper Scientific
Project Timer, Sper Scientific
2 Options Available Below
Product Details & Documents
This long range digital countdown project timer allows you to set a target day for completion of a project up to 8 years in the future and the unique timer counts down to your deadline in days, hours, minutes, seconds and ¹/₁₀₀ seconds on a 12 digit LCD.
  • Standard clock, timer, and stopwatch modes
  • A tripod back enables the project timer to stand on a bench top
View either the target date or the actual count down. You can insert the name of your project into a special window which can be seen on the timer face.
Certifications: N.I.S.T. traceable certificate of compliance for 14003-160.
Ordering information: One AAA battery included.
Product Family Options
Product Information
  • Désignation
    W×H×D
    Weight
    Availability
    Price
  • Project timer
    130×64×20 mm (5×2⅞×⅞")
    71 g (2.5 oz.)
    Log in for availability
    Log in for availability
    Each
    Each - $73.97
    $73.97
    Each
       $73.97
    Supplier Number: 810017
  • Project timer, certified
    130×64×20 mm (5×2⅞×⅞")
    71 g (2.5 oz.)
    Log in for availability
    Log in for availability
    Each
    Each - $79.38
    $79.38
    Each
       $79.38
    Supplier Number: 810017C