Silicone Safety Pipette Filler, Sper Scientific
Pipette fillers enable filling and dispensing of pipettes with one hand.
- Adjusts to fit any size pipet
- Chemically resistant glass valves
- Available in two colors for identification purposes
Safety is ensured through avoidance of hand or mouth contact with liquid. Our Silicone Pipette Fillers offer a softer feel with easier and faster operation of valves. Silicone is also more chemically resistant and more resilient than either PVC or natural rubber. It will not crack or become brittle and will last longer and stay flexible.
: Glass tube not included, order separately.
Frequently Bought Together