Lab Digital Salinity Refractometer, Sper Scientific | Avantor
Lab Digital Salinity Refractometer, Sper Scientific
Lab Digital Salinity Refractometer, 180 x 100 x 65 mm (7 x 4 x 21/2"), 500 g (17.6 oz.)
Catalog # 97011-860
Supplier:   Sper Scientific
Fournisseur #:  300035
Lab Digital Salinity Refractometer, Sper Scientific
Catalog # 97011-860
Supplier:  Sper Scientific
Lab Digital Salinity Refractometer, Sper Scientific
Lab Digital Salinity Refractometer, Sper Scientific
Lab Digital Salinity Refractometer, 180 x 100 x 65 mm (7 x 4 x 21/2"), 500 g (17.6 oz.)
Price
$1,511.01
Log in pour la disponibilité
Each
Product Details & Documents
Measure concentrations in brix, salinity, refractive index, and clinical parameters with the full scale accuracy of 0.1% using CCD scanner (linear scanned array imaging) technology
These simple to use models calibrate to distilled water and require a sample of only 0.4mL. Instantly displays temperature in addition to multiple scales with the unit of measure indicated on the large LCD. The sophisticated microprocessor provides automatic calibration, automatic temperature compensation, automatic shutoff (after 3 minutes), and visual min./max. alarms. Low power consumption enables 5,000 readings, before the LCD indicates that the battery should be replaced. The water-resistant design protects the electronics from wet environments and allows for easy clean ups.
Ordering information: Supplied ready to use with a 9V battery, distilled water, transfer-pipet, instructions, and a protective case. The refractometer also includes a 1-year warranty.
Spécifications
  • Désignation:
    Lab Digital Salinity Refractometer
  • Dimensions:
    180 x 100 x 65 mm (7 x 4 x 21/2")
  • Weight:
    500 g (17.6 oz.)