Impinger, Air Sampling, High Velocity, Ace Glass
Fournisseur: Ace Glass
Certificats
À propos de cet article
This high velocity impinger passes 6L/min
(corrected to engineering standard gas conditions) when the pressure drop across the orifice is 41cm Hg. or greater. The tip of the capillary stem is 4mm from the flask bottom. Joint is [ST] 24/25. Center tube is 11mm O.D.; side tube is 8mm O.D. with bulge. Fabricated from borosilicate glass.
Ordering information: Complete item consists of 89083-006 bottle and 89059-374 tube. Components may also be purchased separately.
Frequently Bought Together