Order Entry
Canada
ContactUsLinkComponent
 
Human C-Type Natriuretic Peptide (1-53) Trifluoroacetate
 
Human C-Type Natriuretic Peptide (1-53) Trifluoroacetate

 

  • CAS No.:
    141294-77-1
  • Species:
    Human
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00211819
  • Protein synonyms:
    CNP-53
  • Protein/peptide name:
    C-Type Natriuretic Peptide (1-53) Trifluoroacetate
  • Molecular weight:
    5801.77
  • Sequence:
    H-Asp-Leu-Arg-Val-Asp-Thr-Lys-Ser-Arg-Ala-Ala-Trp-Ala-Arg-Leu-Leu-Gln-Glu-His-Pro-Asn-Ala-Arg-Lys-Tyr-Lys-Gly-Ala-Asn-Lys-Lys-Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys-OH
  • Molecular formula:
    C₂₅₁H₄₁₇N₈₁O₇₁S₃

 

Frequently Bought Together