H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH Trifluoroacetate
Fournisseur: Bachem Americas
Certificats
À propos de cet article
The integrin-binding heptapeptideGRGDSPC can be easily linked to carriers or surfaces.
Spécifications
- CAS No.:91575-26-7
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00076461
- Protein synonyms:GRGDSPC
- Protein/peptide name:H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH Trifluoroacetate
- Molecular weight:690.74
- Sequence:H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH
- Molecular formula:C₂₅H₄₂N₁₀O₁₁S