Eel (Des-Cys¹,cyclo(Ser²-Asu⁷))-Calcitonin Acetate
Fournisseur: Bachem Americas
Certificats
À propos de cet article
For Elcatonin see H-2214.
Elcatonin, also known as carbocalcitonin, is the aminosuberic acid analog of eel calcitonin. It has all the biological properties of the corresponding natural calcitonin (H-2255). The substitution of the disulfide bond of natural calcitonins with an ethylene bridge in 1-7 N-terminal position gives elcatonin greater stability and excellent tolerability when used in vivo.
Spécifications
- CAS No.:60731-46-6
- Species:Eel
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD10566642
- Protein synonyms:Elcatonin|(Asu1·7)-Calcitonin (eel)
- Protein/peptide name:(Des-Cys¹,cyclo(Ser²-Asu⁷))-Calcitonin Acetate
- Molecular weight:3363.82
- Sequence:Cyclo(-Ser-Asn-Leu-Ser-Thr-Asu)-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asp-Val-Gly-Ala-Gly-Thr-Pro-NH₂
- Molecular formula:C₁₄₈H₂₄₄N₄₂O₄₇