Cyclo(-Pro-Tyr)
Cyclo(-Pro-Tyr)
Supplier:   Bachem Americas
Cyclo(-Pro-Tyr)
Supplier:  Bachem Americas
Certificates
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
This host-specific phytotoxin produced by Alternaria alternata on spotted knapweed is an ideal prototype for creating a safe and an environmentally friendly antiknapweed herbicide. Furthermore, this cyclic dipeptide induced a high degree of phosphorylation of histones.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • G-4715.0250BA
  • G-4715.1000BA
  • Protein/peptide name:
  • Cyclo(-Pro-Tyr)
  • Cyclo(-Pro-Tyr)
  • Protein synonyms:
  • Maculosin
  • Maculosin
  • CAS No.:
  • 4549-02-4
  • 4549-02-4
  • MDL No.:
  • MFCD03093467
  • MFCD03093467
  • Sequence:
  • Cyclo(-Pro-Tyr)
  • Cyclo(-Pro-Tyr)
  • Molecular weight:
  • 260.29
  • 260.29
  • Molecular formula:
  • C₁₄H₁₆N₂O₃
  • C₁₄H₁₆N₂O₃
  • Storage conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C
Specifications
Product Family Options
Product Information
  • Taille
    Availability
    Price
  • 250 mg
    Log in for availability
    Log in for availability
    Each (250mg)
    Each (250mg) - $219.08
    $219.08
    Each (250mg)
       $219.08
    Minimum order quantity is 1
    Supplier Number: 4029436.0250
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    G-4715.0250BA
    Protein/peptide name
    Cyclo(-Pro-Tyr)
    Protein synonyms
    Maculosin
    CAS No.
    4549-02-4
    MDL No.
    MFCD03093467
    Sequence
    Cyclo(-Pro-Tyr)
    Molecular weight
    260.29
    Molecular formula
    C₁₄H₁₆N₂O₃
    Storage conditions
    –20 ± 5 °C
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $641.29
    $641.29
    Each (1g)
       $641.29
    Minimum order quantity is 1
    Supplier Number: 4029436.1000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    G-4715.1000BA
    Protein/peptide name
    Cyclo(-Pro-Tyr)
    Protein synonyms
    Maculosin
    CAS No.
    4549-02-4
    MDL No.
    MFCD03093467
    Sequence
    Cyclo(-Pro-Tyr)
    Molecular weight
    260.29
    Molecular formula
    C₁₄H₁₆N₂O₃
    Storage conditions
    –20 ± 5 °C