Order Entry
Canada
ContactUsLinkComponent
H-Leu-Ala-OH
Fournisseur:  Bachem Americas
Certificats
H-Leu-Ala-OH
Fournisseur:  Bachem Americas

Spécifications

  • CAS No.:
    7298-84-2
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00002641
  • Protein/peptide name:
    H-Leu-Ala-OH
  • Molecular weight:
    202.25
  • Sequence:
    H-Leu-Ala-OH
  • Molecular formula:
    C₉H₁₈N₂O₃

Spécifications

Frequently Bought Together