H-Ile-Val-OH
Catalogue # G-2445.0001BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:41017-96-3
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038332
- Protein/peptide name:H-Ile-Val-OH
- Molecular weight:230.31
- Sequence:H-Ile-Val-OH
- Molecular formula:C₁₁H₂₂N₂O₃
- Cat. no.:G-2445.0001BA
- Supplier No.:G-2445.0001
Spécifications
À propos de cet article
An effector dipeptide that enhances the catalytic activity of the proenzyme trypsinogen. Forms nanotubes as its retroanalog H-Val-Ile-OH. These pores absorb and store gases as methane or hydrogen.