Cyclo(-Phe-Ser)
1 g
Catalog # G-1770.0001BA
Supplier: Bachem Americas
Fournisseur #: 4004737.0001
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
An intermediate in the biosynthesis of gliotoxin.
Spécifications
Spécifications
- Taille:1 g
- Cat. No.:G-1770.0001BA
- Molecular formula:C₁₂H₁₄N₂O₃
- Protein/peptide name:Cyclo(-Phe-Ser)
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00056023
- Sequence:Cyclo(-Phe-Ser)
- Molecular weight:234.26
- CAS No.:35591-00-5



