Cyclo(-His-Phe)
À propos de cet article
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
Catalyst for asymmetric cyanohydrin synthesis. The addition of hydrogen cyanide to benzaldehyde gives the highest optical yield ever obtained for the corresponding cyanohydrin. In a study of McCleland et al., G-1740 showed significant anti-tumour activity, causing reduction of cell viability in cervical cancer cells. In isolated rat hearts, the diketopiperazine caused a gradual reduction of heart rate and a decrease in coronary flow rate.
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
Catalyst for asymmetric cyanohydrin synthesis. The addition of hydrogen cyanide to benzaldehyde gives the highest optical yield ever obtained for the corresponding cyanohydrin. In a study of McCleland et al., G-1740 showed significant anti-tumour activity, causing reduction of cell viability in cervical cancer cells. In isolated rat hearts, the diketopiperazine caused a gradual reduction of heart rate and a decrease in coronary flow rate.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- G-1740.0250BA
- G-1740.1000BA
- Protein/peptide name:
- Cyclo(-His-Phe)
- Cyclo(-His-Phe)
- CAS No.:
- 56586-95-9
- 56586-95-9
- MDL No.:
- MFCD00135317
- MFCD00135317
- Sequence:
- Cyclo(-His-Phe)
- Cyclo(-His-Phe)
- Molecular weight:
- 284.32
- 284.32
- Molecular formula:
- C₁₅H₁₆N₄O₂
- C₁₅H₁₆N₄O₂
- Storage conditions:
- –20 ± 5 °C
- –20 ± 5 °C
Spécifications
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- Supplier Number: 4006752.0250L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.G-1740.0250BAProtein/peptide nameCyclo(-His-Phe)CAS No.56586-95-9MDL No.MFCD00135317SequenceCyclo(-His-Phe)Molecular weight284.32Molecular formulaC₁₅H₁₆N₄O₂Storage conditions–20 ± 5 °C - Supplier Number: 4006752.1000L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.G-1740.1000BAProtein/peptide nameCyclo(-His-Phe)CAS No.56586-95-9MDL No.MFCD00135317SequenceCyclo(-His-Phe)Molecular weight284.32Molecular formulaC₁₅H₁₆N₄O₂Storage conditions–20 ± 5 °C