CulGenex™ Heat-and-Pour™ Agarose Gel Formulations & Reagents, Hardy Diagnostics
Catalog # CA89407-062
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:PC bottle
- Description:LB Agar (Heat in microwave and pour), 1000 ml polycarbonate bottle, 500 ml fill
- Size:500 ml
- Storage temperature:2-30C
- Minimum Order Quantity:4
- Cat. no.:CA89407-062
- Final Volume:1000 ml
- Supplier No.:CG60BX
Specifications
About this item
CulGenex™ Heat-and-Pour™ Agarose Gels, which contain ethidium bromide, agarose, and with either TAE or TBE buffers, are recommended for agarose gel electrophoresis in the separation, visualization, quantification, or isolation of particular DNA fragments.
Just microwave and pour.
LB Agar Media formulations are recommended for the cultivation of bacteria used in molecular biology studies.
LB Agar Media formulations are recommended for the cultivation of bacteria used in molecular biology studies.