pH Electrode Storage Solution, Hach
pH Electrode Storage Solution, Hach
pH Electrode Storage Solution, 500 mL Bottle
Catalog # CA27565-49
Supplier:   Hach
Supplier #:  2756549
pH Electrode Storage Solution, Hach
Catalog # CA27565-49
Supplier:  Hach
CAS Number:  
pH Electrode Storage Solution, Hach
pH Electrode Storage Solution, Hach
pH Electrode Storage Solution, 500 mL Bottle
CAS Number:  
Price
$122.44
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Potassium Chloride (KCl) 3 M.
Specifications
Specifications
  • Désignation:
    pH Electrode Storage Solution
  • Taille:
    500 mL Bottle