BOD Nutrient Buffer Pillows, 300 mL, Hach
BOD Nutrient Buffer Pillows, 50 pillows, 0.5 ml, 300 ml
Catalog # CA14160-66
Supplier:   Hach
 :  1416066
BOD Nutrient Buffer Pillows, 300 mL, Hach
Catalog # CA14160-66
Supplier:  Hach
CAS Number:  
BOD Nutrient Buffer Pillows, 300 mL, Hach
BOD Nutrient Buffer Pillows, 300 mL, Hach
BOD Nutrient Buffer Pillows, 50 pillows, 0.5 ml, 300 ml
CAS Number:  
Price
$41.50
Log in pour la disponibilité
Pack of 50 (0.5.ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For preparation of BOD5 dilution water
Ready for immediate use. 0.5-mL size powder pillows. Each pillow prepares 300 mL of dilution water.
Ordering information: Pack of 50 pillows.
Spécifications
Spécifications
  • Désignation:
    BOD Nutrient Buffer Pillows
  • Taille:
    50 pillows
  • Final Volume:
    300 ml
  • Volume:
    0.5 ml