Citric Acid F Reagent Solution, Hach
Catalogue # CA22542-49
Fournisseur: Hach
Numéro CAS:
Spécifications
- Chemical method:Heteropoly blue
- Description:Citric Acid F Reagent Solution
- Size:500 mL bottle
- Method:8282 - Heteropoly Blue, Rapid Liquid
- Cat. no.:CA22542-49
- Range:3-1000 μg/L SiO₂
- Supplier No.:22542-49
- No. of Tests:250
Spécifications
À propos de cet article
For determination of ultra-low range silica by the Heteropoly Blue method (Hach Method 8282)
Range: 3-1000 μg/L SiO2.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.