Hardness 1 Buffer Solution, 100 mL MDB, Hach
Hardness 1 Buffer Solution, 100 mL MDB, Hach
Supplier:   Hach
Hardness 1 Buffer Solution, 100 mL MDB, Hach
Supplier:  Hach
Certificates
Hardness 1 Buffer Solution, 100 mL MDB, Hach
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C
Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA424-32
  • CA424-49
  • CA42453
  • Method:
  • 8226, 8213; EDTA Titration
  • 8213, 8226 - EDTA Titration
  • EDTA Titration
Specifications
Product Family Options
Product Information
  • Désignation
    Taille
    Méthode chimique
    Type de conditionnement
    Availability
    Price
  • Hardness 1 Buffer Solution, 100 mL MDB
    100 ml
    EDTA
    Marked dropping bottle (MDB)
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $56.71
    $56.71
    Each (100ml)
       $56.71
    Supplier Number: 42432
    Specifications:
    More
    Specifications
    Cat. No.
    CA424-32
    Method
    8226, 8213; EDTA Titration
  • Hardness 1 Buffer Solution
    500 ml
    EDTA
    Bottle
    Log in for availability
    Log in for availability
    Each
    Each - $105.15
    $105.15
    Each
       $105.15
    Supplier Number: 42449
    Specifications:
    More
    Specifications
    Cat. No.
    CA424-49
    Method
    8213, 8226 - EDTA Titration
  • Hardness 1 Buffer Solution
    1 L
    EDTA
    Bottle
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $199.15
    $199.15
    Each (1l)
       $199.15
    Supplier Number: 42453
    Specifications:
    More
    Specifications
    Cat. No.
    CA42453
    Method
    EDTA Titration