Stablcal® Turbidity Standards Calibration Kit, 2100Q Portable Turbidimeter, Hach
Catalog # CA11014-240
Supplier: Hach
CAS Number:
Specifications
- Turbidity:100 NTU800 NTU10 NTU20 NTU
- Description:Stablcal® Turbidity Standards Calibration Kit, Sealed Vials
- Size:4 Ampule
- Concentration:20, 100, 800, 10 NTU (for verification)
- Cat. no.:CA11014-240
- For Use With:2100Q Portable Turbidimeter
- Supplier No.:2971205
Specifications
About this item
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter
- Precise, Pre-mixed Formazin Solutions
- Based on Formazin Primary Standards
- Enhanced Stability Saves Time and Labor
- Confidence in results
Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit contains one each of 20, 100, and 800 NTU Stablcal vials for calibration and one 10 NTU Stablcal vial for verification.