Boss Head, Square
Boss Head - Square
Catalog # 470137-832
Supplier: Wards
Fournisseur #: CH0664
Jump to:
Product Details & Documents
Perfect for Physics
This square boss head accommodates both square and tubular sections up to 16mm and holds them at right angles to each other. Holds a wide variety of objects including: rules, magnets, drawing boards, and more. The alloy body and plastic thumbscrews make it perfect for any physics lab.
Spécifications
- Désignation:Boss Head - Square