Process Pipe Couplings, Bead to Bead, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Couplings consist of a .300 series stainless steel outer band with a T-Bolt and hex nut, a fluoroelastomer cushion liner and a TFE seal ring.
Designed for making beaded end to beaded end connections of process glass drain pipe and can withstand up to 204 °C (400 °F).
Frequently Bought Together