Filter Support Assembly, Ace Glass
À propos de cet article
These filter support assembly are used to collect dust particles in a stack sampling system.
- Available in either 51 mm (2"), 76 mm (3"), or 102 mm (4") sizes
The filter assembly part A consists of [SJ] 28/15 outer joint bent perpendicular to flow path and part B consists of a seat for a fritted disc and a [SJ] 28/15 inner joint. Likewise, part C consists of a neoprene gasket used to seal filter paper and prevent leakage around the filter and part D with a Porosity B fritted disc.
: Positive, two-piece aluminum clamps feature a gaskets and brass tightening bolts for securing the 76 mm (89059-262) or 102 mm (89059-264) size filter support assembly. The bolt head is flanged to secure the top half as the bolt is threaded into the bottom half.
: Parts A, B, C, and D must be purchased separately. Two-piece clamps need to be ordered separately.
12 Options Available Below
Product Details & Documents
These filter support assembly are used to collect dust particles in a stack sampling system.
- Available in either 51 mm (2"), 76 mm (3"), or 102 mm (4") sizes
The filter assembly part A consists of [SJ] 28/15 outer joint bent perpendicular to flow path and part B consists of a seat for a fritted disc and a [SJ] 28/15 inner joint. Likewise, part C consists of a neoprene gasket used to seal filter paper and prevent leakage around the filter and part D with a Porosity B fritted disc.
: Positive, two-piece aluminum clamps feature a gaskets and brass tightening bolts for securing the 76 mm (89059-262) or 102 mm (89059-264) size filter support assembly. The bolt head is flanged to secure the top half as the bolt is threaded into the bottom half.
: Parts A, B, C, and D must be purchased separately. Two-piece clamps need to be ordered separately.
Product Family Options
Informations sur le produit
- DésignationFor Use WithClamp SizeAvailabilityPrice
- Supplier Number: 7519-04
- Supplier Number: 7519-22
- Supplier Number: 7519-08
- Supplier Number: 7519-11
- Supplier Number: 7519-05
- Supplier Number: 7519-25
- Supplier Number: 7519-09
- Supplier Number: 7519-12
- Filter Support Assembly, Part AClamp 89055-044102 mmLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,073.88$1,073.88Prix promotionnel pour les nouveaux clientsEach$1,073.88Supplier Number: 7519-06
- Supplier Number: 7519-30
- Supplier Number: 7519-10
- Supplier Number: 7519-13
Accessories
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Two Piece Clamp, Flat Flange, 160×120 mmTwo Piece Clamp, Flat Flange, 160×120 mm89055-044Filter Support Assembly, 102 mmLog in pour la disponibilitéEach - $501.91
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Two Piece Clamp, Flat Flange, 152×111 mmTwo Piece Clamp, Flat Flange, 152×111 mm89055-042Filter Support Assembly, 76 mmLog in pour la disponibilitéEach - $532.54