Flask, Two-Neck, Pear-Shaped, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
These pear-shaped flasks have a side arm for use with a rubber stopper (not included)
The center joint is [ST] 14/20 outer. Fabricated from heavy wall borosilicate glass -- approximately 30% thicker than standard wall flasks.
Frequently Bought Together