Flask with Thermowell, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
This round-bottom flask has a center neck with a [SJ] 35/25 top joint and an angled side port thermometer well
Fabricated from borosilicate glass.
Frequently Bought Together