Flask Reaction, Cylindrical, with Zero Dead Space Valve, Ace Glass Incorporated
This cylindrical reaction flask has a Duran style top flange with an o-ring groove for use with matching heads such as 89054-372, 89055-320, or 89055-326 (among others)
The bottom outlet is a Zero Dead Space (ZDS™) PTFE valve with Chemraz o-rings. This flask has heavy walls that are 30% thicker than standard wall flasks. Fabricated from borosilicate glass.
: Comes complete with one CAPFE o-ring (89061-166 for 100mm flanges, 89061-168 for 150mm flanges). For optional silicone o-rings, order 71000-162 (100mm) or 71000-164 (150mm). Use a 89055-110 (100mm) or 89055-112 (150mm) clamp to secure head to flask.
Frequently Bought Together