Reaction Flask with Detached Stopcock, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
This reaction flask has a detached 4mm bore glass stopcock
The flask is separated from the stopcock and outlet tube by a [SJ] 28/15 joint, ball on the flask, socket on the stopcock. Fabricated from borosilicate glass.
Ordering information: Complete apparatus includes flask and stopcock. For bottom joint clamp (not included), order 89060-256. For a clamp (not included) to secure the 4" head of your choice (not included) to the flask, order 89055-014.
Frequently Bought Together