Reaction Flask with Indents, 4-Inch Conical Flange, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
This rugged, cylindrical reaction flask has a conical neck opening of four inches (100 mm)
It has side indents for improved stirring characteristics. Not suitable for vacuum operation. Use a 89055-014 clamp to secure the head to the flask. For gaskets, see 89055-010 (or similar). Fabricated from borosilicate glass.
Ordering information: Clamp and gasket not included. Use a 89055-014 clamp to secure the head to the flask. For gaskets, see 89055-004.
Frequently Bought Together