Corning® MCDB 131 1X, Corning
Corning® MCDB 131 1X, Corning
MCDB 131 1X
Catalog # CA45001-112
Supplier:   Corning
Supplier #:  15-100-CV
Corning® MCDB 131 1X, Corning
Catalog # CA45001-112
Supplier:  Corning
CAS Number:  
Corning® MCDB 131 1X, Corning
Corning® MCDB 131 1X, Corning
MCDB 131 1X
CAS Number:  
Price
$426.00
Log in pour la disponibilité
Pack of 6
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
MCDB 131 was originally developed by the researchers Knedler and Ham for the low-protein and serum free growth of human microvascular endothelial cells (HMVEC)
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Since its development, this medium has been used for this cell type and others such as liver, smooth muscle, cardiac myocytes. It is one of a group of MCDB media which were designed to provide a defined and optimally balanced nutritional environment for various mammalian cell types.
Spécifications
Spécifications
  • Désignation:
    MCDB 131 1X
  • Cat. No.:
    CA45001-112
  • Storage temperature:
    Refrigerate
Spécifications