ContactUsLinkComponent
Canister for SATWipes® Presaturated Wipers, Contec
Catalogue # 68311-030
Fournisseur:  Contec
Small Business Enterprise
undefined
Canister for SATWipes® Presaturated Wipers, Contec
Catalogue # 68311-030
Fournisseur:  Contec
Supplier Number:  SAT-CAN120
Small Business Enterprise
Spécifications
  • Description:
    Canister with snap-close lid, unfilled, for use with SATWipes 6 x 9"/100 count rolls, 10 canisters/case
  • Cat. no.:
    68311-030
  • Supplier No.:
    SAT-CAN120
Spécifications
À propos de cet article
This reusable canister holds pre-saturated wipes that are 6" x 9" (152×229 mm) and are dispensed from center-pull rolls.
Canister comes with a snap close lid that helps keep wipes saturated and clean.