KLEENEX® Hand Towels in a POP-UP™ Box, KIMBERLY-CLARK PROFESSIONAL®
Catalogue # CA11007-732
Fournisseur: Kimberly-Clark
Spécifications
- Color:White
- Description:Hand Towels in a POP-UP™ Box
- Cat. no.:CA11007-732
- Supplier No.:01701
Spécifications
À propos de cet article
Say goodbye to soggy, messy stacks of paper hand towels.
- More convenient, hygienic countertop towel solution
- Water-resistant coating prevents the box from getting wet
KLEENEX® POP-UP™ box hand towels provide hygienic one-at-a-time dispensing, since each person touches only the towel they use. Easy maintenance, since there is no dispenser upkeep or time-consuming reloading process.
They're a great choice for high-end offices, lodging, or any facility that prefers countertop dispensing. Meets EPA Standards for minimum post consumer waste content: towels 40%. 50% recycled fiber content. FSC & EcoLogo Certified.