ContactUsLinkComponent
JACKSON SAFETY® Lens Cleaning Towelettes, KIMBERLY-CLARK PROFESSIONAL®
Catalog # 33300-562
Supplier:  Ansell Healthcare
undefined
JACKSON SAFETY® Lens Cleaning Towelettes, KIMBERLY-CLARK PROFESSIONAL®
Catalog # 33300-562
Supplier:  Ansell Healthcare
Supplier Number:  14551
Specifications
  • Description:
    Lens cleaning towelettes
  • Cat. no.:
    33300-562
  • Supplier No.:
    14551
Specifications
About this item
Anti-fog, anti-static formula for cleaning glass, plastic, and polycarbonate lenses, goggles, and visors.
Packaging: 100 individually packaged towelettes per box.