SCOTT® Personal Seats, KIMBERLY-CLARK PROFESSIONAL®
CA21908-693
- Color:White
- Description:Personal Flushable Sanitary Seats, 1-Ply
- Size:14.5 x 17" Sheets
- Cat. no.:CA21908-693
- Supplier No.:39000
SCOTT® toilet seat covers are flushable and offer sanitary protection, and help to reduce litter and clogging.
: Meets EPA guidelines for post consumer waste: bathroom tissue 20%; 20% post-consumer waste; 50% recycled fiber.