Large Evaporator Flasks, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
These large evaporator flasks are used with Heidolph 20L rotary evaporators
They are fabricated from blanks selected for balance and quality. The necks are carefully welded to prevent “rotational whip.” Fabricated from borosilicate glass. Flange A O.D. is 143mm. Flange B O.D. is 118mm. Flange C O.D. is 18mm.
Frequently Bought Together