Alkaline running buffer solution, Thermo Scientific
Alkaline running buffer solution, Thermo Scientific
Alkaline running buffer solution, Thermo Scientific
Certificates
Alkaline running buffer solution, Thermo Scientific
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CAAAJ63170-AK
  • CAAAJ63170-AP
  • Physical form:
  • Liquid
  • Liquid
Specifications
Product Family Options
Product Information
  • Taille
    Availability
    Price
  • 250 mL
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $57.99
    $57.99
    Each (250ml)
       $57.99
    Minimum order quantity is 1
    Supplier Number: J63170-AK
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ63170-AK
    Physical form
    Liquid
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $102.26
    $102.26
    Each (500ml)
       $102.26
    Minimum order quantity is 1
    Supplier Number: J63170-AP
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ63170-AP
    Physical form
    Liquid