Wash buffer, pH 8.0, Thermo Scientific
CAAAJ61918-AP
:
- Size:500 mL
- Cat. no.:CAAAJ61918-AP
- Supplier No.:J61918-AP
Liquid
Physical Form: Liquid
Applications: For His-tag protein purification
Liquid
Physical Form: Liquid
Applications: For His-tag protein purification