Succinate, 0.2 M Buffer Solution, pH 6.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Succinate, 0.2 M Buffer Solution, pH 6.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 150-90-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00002789
2 Options Available Below
Product Details & Documents
Liquid
CAS No.: 150-90-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00002789
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CAAAJ61853-AK
- CAAAJ61853-AP
- MDL No.:
- MFCD00002789
- MFCD00002789
Specifications
Product Family Options
Product Information
- TailleAvailabilityPrice
- Supplier Number: J61853-AKSpecifications:
More
Specifications
Cat. No.CAAAJ61853-AKMDL No.MFCD00002789 - Supplier Number: J61853-APSpecifications:
More
Specifications
Cat. No.CAAAJ61853-APMDL No.MFCD00002789



