Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific
Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific
Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific
Certificates
Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific
2 Options Available Below
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Blue
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 25 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (25ml)
    Each (25ml) - $125.66
    $125.66
    Each (25ml)
       $125.66
    Minimum order quantity is 1
    Supplier Number: J61042-AC
  • 50 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (50ml)
    Each (50ml) - $110.48
    $110.48
    Each (50ml)
       $110.48
    Minimum order quantity is 1
    Supplier Number: J61042-AD