Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

CHAPS Lysis Buffer (2X), Thermo Scientific

CHAPS Lysis Buffer (2X), Thermo Scientific
Certificates

1 Options Available Below

Product Details & Documents

Liquid

Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866

Specifications for Product Family

Spécifications

Spécifications

  • Catalog No:
  • CAAAJ60812-AC
  • CAAAJ60812-AD
  • MDL No.:
  • MFCD00282866
  • MFCD00282866

Spécifications

Product Family Options

Informations sur le produit

  • Taille
    Availability
    Price
  • 25 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (25ml)
    Each (25ml) - $207.11
    $207.11
    Each (25ml)
       $207.11
     

    Supplier Number: J60812-AC
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAAJ60812-AC
    MDL No.
    MFCD00282866
  • 50 mL
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAAJ60812-AD
    MDL No.
    MFCD00282866
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.