DNAzol® Reagent, Thermo Scientific
DNAzol® Reagent, Thermo Scientific
DNAzol® Reagent, Thermo Scientific
Certificates
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DNAzol® Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin.
  • Liquid, soluble in water
  • Store at ambient temperature
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • CAAAJ60637-AD
  • MPCAAAJ60637-AE
  • CAAAJ60637-AE
  • CAAAJ60637-AH
  • Target molecule:
  • DNA
  • DNA
  • DNA
  • DNA
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • DNAzol® Reagent
    50 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (50ml)
    Each (50ml) - $408.22
    $408.22
    Each (50ml)
       $408.22
    Supplier Number: J60637-AD
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAAJ60637-AD
    Target molecule
    DNA
  • DNAzol® Reagent
    100 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100ml)
    Each (100ml) - $746.73
    $746.73
    Each (100ml)
       $746.73
    Supplier Number: J60637-AE
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAAJ60637-AE
    Target molecule
    DNA
  • DNAzol® Reagent
    200 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (200ml)
    Each (200ml) - $1,390.56
    $1,390.56
    Each (200ml)
       $1,390.56
    Minimum order quantity is 1
    Supplier Number: J60637-AH
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAAJ60637-AH
    Target molecule
    DNA
Life Science Promotion
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • DNAzol® Reagent
    100 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $462.00
    $462.00
    Each
       $462.00
    Supplier Number: J60637-AE
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    MPCAAAJ60637-AE
    Target molecule
    DNA