Order Entry
Canada
ContactUsLinkComponent
TPE (10X), Thermo Scientific
TPE (10X), Thermo Scientific
Certificates
TPE (10X), Thermo Scientific
Certificates

Specifications

  • MDL No.:
    MFCD00132488
  • Storage temperature:
    RT
  • Physical form:
    Liquid

Specifications

Frequently Bought Together