ContactUsLinkComponent
Top Loader Workstation, PVC, TrippNT
  11376-290
 :  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Top Loader Workstation, PVC, TrippNT
  11376-290
 :  TrippNT
 :  50027
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • Description:
    Top Loading Workstation with 8 Compartments and 6 Slot Tool Holder
  • Cat. no.:
    11376-290
  • W×D×H:
    33×20×30 cm (13×8×12")
  • Weight:
    1 kg (3 lbs.)
  • Supplier No.:
    50027
White PVC top loader workstation has all the features you need to consolidate and stream-line your processes.
Made from durable white PVC plastic this workstation includes eight flexible compartments to organize tools like pliers, spatulas, hemostats, or other tools. There is also plenty of room for 6" weigh papers, 6" boats, calculators, and clipboards. It also has a built in holder for a small box of Kimwipes® or other wipers.