Equipment Draft Shields, Clear Acrylic, TrippNT
Fournisseur: TrippNT
Certificats
À propos de cet article
Crystal clear acrylic draft shield protects balances and delicate instruments from air drafts and movement.
- Cube-designed shield with draft deflecting overhang
- Made from clear ³/₁₆" acrylic
- Blocks air drafts around delicate equipment
- 100% total replacement lifetime guarantee
- Made in USA
Shields are completely see-through, with a half-circle cutout in the back panel for threading cords or wires. Shield front features a 7.6 cm (3") downdraft guard the full length of the shield. Installs by simply placing it over the equipment requiring protection. One side is open for easy access.
Available in four sizes; 13, 15, 18 and 22" cubes.
Frequently Bought Together