Hexanamide ≥98.0%
Hexanamide ≥98.0%
Hexanamide ≥98.0%
Supplier:  TCI America
Certificates
Hexanamide ≥98.0%
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CAS Number: 628-02-4
MDL Number: MFCD00008046
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 100
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Capronamide
Synonymes:  Capronamide
  • CAS Number:628-02-4
  • Formule:C₆H₁₃NO
  • Numéro MDL:MFCD00008046
  • Molecular Weight:115.18 g/mol
  • Fusion Pt:100 °C
Specifications for Product Family
Résultats des tests de spécification
  • Purity/Analysis Method:
    98.0% (N)
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5g)
    Each (5g) - $64.50
    $64.50
    Each (5g)
       $64.50
    Supplier Number: C0024-5G
  • 25 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (25g)
    Each (25g) - $187.00
    $187.00
    Each (25g)
       $187.00
    Supplier Number: C0024-25G