STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe® | Avantor
STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe®
Supplier:   ITW Texwipe
STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe®
Supplier:  ITW Texwipe
Certificates
STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe®
STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe®
1 Options Available Below
Product Details & Documents
Sterile AlphaSat® incorporates Texwipe's highly sorptive AlphaSorb® HC polyester wipers plus 70% IPA.
  • Constructed from a unique two-ply, pinsonic fabric
  • 100% knitted continous-filament polyester
Chosen for their low particle-generating characteristics and low extractable levels, these wipers offer superior strength in a continous-filament knitted substrate.
This wiper is saturated with a 0.2 μm filtered, 70% isopropyl alcohol/30% deionized water solution. Packaging permits alcohol wipedown of exterior bag before introduction of the inner bag into sterile suites. Gamma irradiated to 10⁻⁶ Sterility Assurance Level. Fibers (>100 μm) are 230 fibers/m². The isopropyl alcohol extractant is 0.09 g/m². The deionized water extractant is 0.05 g/m².
Sterile AlphaSat® with AlphaSorb® HC is ideal for alcohol wipedowns in the most critical areas of pharmaceutical sterile environments.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CATWTX3252
  • TWTX3252
  • Basis weight:
  • 195 g/m²
  • 195 g/m²
  • Absorbency:
  • 595 ml/m²
  • 595 ml/m²
  • Absorbency rate:
  • 0.3 s
  • 0.3 s
  • Particle generation (0,5 - 5 µm) - ASTM E2090 test:
  • 6 × 10⁶/m²
  • 6 × 10⁶/m²
  • Particle generation (5 - 100 µm) - ASTM E2090 Test:
  • 315,000/m²
  • 315,000/m²
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Sterility
    Packaging
    Availability
    Price
  • Wipes with 70% IPA
    12 x 12"
    Gamma irradiated
    25 wipers/flatpack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $345.00
    $345.00
    Log in pour la disponibilité
    Case of 125
    Case of 125 - $1,725.00
    $1,725.00
    Pack of 25
       $345.00
    Case of 125
       $1,725.00
    Supplier Number: TX3252
    Spécifications:
    More
    Specifications
    Cat. No.
    CATWTX3252
    Basis weight
    195 g/m²
    Absorbency
    595 ml/m²
    Absorbency rate
    0.3 s
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    6 × 10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    315,000/m²
  • Wipes with 70% IPA
    12 x 12"
    Gamma irradiated
    25 wipers/flatpack
    Discontinued
    Supplier Number: TX3252
    Spécifications:
    More
    Specifications
    Cat. No.
    TWTX3252
    Basis weight
    195 g/m²
    Absorbency
    595 ml/m²
    Absorbency rate
    0.3 s
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    6 × 10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    315,000/m²