ContactUsLinkComponent
Multiple Polypropylene Bucket System, 5 Gallon (20 L), Contec®
Catalogue # 89198-840
Fournisseur:  Contec
Small Business Enterprise
undefined
Multiple Polypropylene Bucket System, 5 Gallon (20 L), Contec®
Catalogue # 89198-840
Fournisseur:  Contec
Supplier Number:  2757
Small Business Enterprise
Spécifications
  • Color:
    Red/Gray
  • Description:
    Compact Stainless Steel and Polypropylene Double Bucket System with Drip Cap and Casters
  • Size:
    20 l (5.3 gal.)
  • Cat. no.:
    89198-840
  • Supplier No.:
    2757
Spécifications
À propos de cet article
The 5 Gallon Multiple Polypropylene Bucket System features a compact electropolished stainless steel rolling cart with two autoclavable polypropylene buckets. A third bucket is available for use when a three bucket system is required.
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Compatible with all Contec mops, depending upon wringer choice
  • System can be used with two or three buckets
  • Handle can be removed for storage
  • Compatible with most disinfectants
Compact design improves maneuverability and reduces storage space. The durable yet lightweight cart has four casters, two that lock easily with the touch of a foot. An extendable frame pulls out from the front of the cart to accommodate the third bucket. Includes a stainless steel connecting kit to prevent drips between the buckets.
The cart features a unique removable handle for easier autoclaving and storage. For maximum flexibility, we offer two types of wringers that are ordered separately. Choose from a stainless steel downpress wringer, or a stainless steel wringer that fits directly on the waste bucket.
Ordering information: Wringer options must be ordered separately. See the Wringers, Contec® product page for compatible options.