Bemcot® M-3 Wipes, Contec®
Bemcot® M-3 Wipes, Quarter-folded, Rayon, 24.9×24.9 cm (10×10"), 100/Bag, 30 Bags/Case
: Contec
Supplier #: M-3
The Bemcot® M3 Wipe features 100% rayon nonwoven fabric. As the wipe only contains rayon, it is extremely absorbent.
- Used extensively in the compact disk and magnetic tape industries
These wipes are quarter folded into a 5×5" square, which is sized to fit the hand and promotes proper wiping protocol.
These soft, low linting, and heat resistant wipes are designed for cleaning high temperature process equipment, diffusion furnaces, and sensitive electronic equipment in cleanroom environments.
- Désignation:Bemcot® M-3 Wipes, Quarter-folded
- Matière:Rayon
- Packaging:100/Bag, 30 Bags/Case
- W×L:24.9×24.9 cm (10×10")