Corning® PureCoat™ ECM Mimetic Flasks, Fibronectin Peptide, Corning
Fournisseur: Corning
Certificats
À propos de cet article
Corning® PureCoat™ ECM Mimetic Flasks, Fibronectin Peptide, Corning® PureCoat™ ECM Mimetic Cultureware is coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent alternative to complex extracellular peptides.
- Animal-free, pre-coated, synthetic peptide surface for predictable scale-up
- Streamlines cell culture workflow, reducing handling and contamination risk
- Even distribution of media across all layers for homogeneous cell growth
- Ability to mix cells and reagents in the Multi-Flask saves time and reduces risk of contamination
- Flexible design lets you pour or aspirate/recover cells using a pipet
Certifications: Manufactured in compliance to cGMP and ISO 9001:2008, ISO 13485:2012 standards.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together