Twill Jean Wipes, Contec
68310-296
: Contec
Small Business Enterprise
- Description:Twill Jean Wipes, Flat Stacked
- Material:Cotton Woven
- Cat. no.:68310-296
- Packaging:300/Bag, 12 Bags/Case
- W×L:22.9×22.9 cm (9×9")
- Supplier No.:TJ-99
The Twill Jean Wipe is a 100% cotton fabric wipe, woven in a 2×1 twill construction.
- Strong and absorbent
- Suitable for high temperature applications
- Available irradiated
- Durable fabric for tough tasks
- Bias cut edges to reduce unraveling
- Sorbent wipe for surface cleaning or spill pick-up
The Twill Jean Wipe is very absorbent and has high thermal stability for high temperature applications. The edges are bias cut to prevent unraveling.